MilliporeSigma
  • Nse1 and Nse4, subunits of the Smc5-Smc6 complex, are involved in Dictyostelium development upon starvation.
Nse1 and Nse4, subunits of the Smc5-Smc6 complex, are involved in Dictyostelium development upon starvation.
Development, growth & differentiation (2015-06-04)
Hideo Taniura, Naoya Tanabe, Yumi Bando, Natsumi Arai
ABSTRACT
The Smc5-Smc6 complex contains a heterodimeric core of two SMC proteins and non-Smc elements (Nse1-6), and plays an important role in DNA repair. We investigated the functional roles of Nse4 and Nse1 in Dictyostelium discoideum. Nse4 and Nse3 expressed as Flag-tagged fusion proteins were highly enriched in nuclei, while Nse1 was localized in whole cells. Using yeast two-hybrid assays, only the interaction between Nse3 and Nse1 was detected among the combinations. However, all of the interactions among these three proteins were recognized by co-immunoprecipitation assay using cell lysates prepared from the cells expressing green fluorescent protein (GFP)- or Flag-tagged fusion proteins. GFP-tagged Nse1, which localized in whole cells, was translocated to nuclei when co-expressed with Flag-tagged Nse3 or Nse4. RNAi-mediated Nse1 and Nse4 knockdown cells (Nse1 KD and Nse4 KD cells) were generated and found to be more sensitive to UV-induced cell death than control cells. Upon starvation, Nse1 and Nse4 KD cells had increases in the number of smaller fruiting bodies that formed on non-nutrient agar plates or aggregates that formed under submerged culture. We found a reduction in the mRNA level of pdsA, in vegetative and 8 h-starved Nse4 KD cells, and pdsA knockdown cells displayed effects similar to Nse4 KD cells. Our results suggest that Nse4 and Nse1 are involved in not only the cellular DNA damage response but also cellular development in D. discoideum.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
PIPES, BioXtra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O