- Effects of microbial phytase on the apparent and standardized total tract digestibility of phosphorus in rice coproducts fed to growing pigs.
Effects of microbial phytase on the apparent and standardized total tract digestibility of phosphorus in rice coproducts fed to growing pigs.
The objectives of this experiment were to determine the apparent total tract digestibility (ATTD) and the standardized total tract digestibility (STTD) of P and the effect of microbial phytase on ATTD and STTD of P in full-fat rice bran (FFRB), defatted rice bran (DFRB), brown rice, broken rice, and rice mill feed when fed to pigs. Ninety-six barrows (initial BW of 19.4 ± 1.4 kg) were allotted to 12 diets with 8 replicate pigs per diet in a randomized complete block design. A basal diet based on corn and soybean meal was formulated. Five additional diets containing corn, soybean meal, and each rice coproduct were also formulated, and the ratio between corn and soybean meal in these diets was similar to that in the basal diet. Six additional diets that were similar to the initial 6 diets with the exception that 1,000 units of microbial phytase were added to the diets were also formulated. The ATTD and STTD of P were calculated for each diet using the direct procedure, and the ATTD and STTD of P in each rice coproduct were calculated using the difference procedure. Results of the experiment indicated that the concentration of P in feces was reduced (P < 0.05) from pigs fed diets with microbial phytase compared with pigs fed diets without phytase. No differences were observed between the basal diet and the broken rice diet, but the ATTD and the STTD of P in those diets was greater (P < 0.05) than in all other diets both without and with phytase. Among the rice coproducts, the greatest (P < 0.05) ATTD and STTD of P were observed for broken rice regardless of inclusion of phytase. If no microbial phytase was used, the values for STTD of P in brown rice, FFRB, DFRB, and rice mill feed were not different, but if microbial phytase was included in the diet, ATTD and STTD of P in brown rice was greater (P < 0.05) than in FFRB, DFRB, and rice mill feed. The STTD of P in brown rice, FFRB, and rice mill feed was greater (P < 0.05) if microbial phytase was used than if no microbial phytase was used. Addition of microbial phytase to the diets also increased (P < 0.05) the ATTD of Ca regardless of the rice coproducts used. In conclusion, the STTD of P is greater in broken rice than in all other rice coproducts. The STTD of P in brown rice, FFRB, DFRB, and rice mill feed is relatively low due to the high concentration of phytate in these ingredients, but addition of microbial phytase will increase the STTD of P in most rice coproducts.