MilliporeSigma
  • Comparison of Mesenchymal Stem Cell Source Differentiation Toward Human Pediatric Aortic Valve Interstitial Cells within 3D Engineered Matrices.
Comparison of Mesenchymal Stem Cell Source Differentiation Toward Human Pediatric Aortic Valve Interstitial Cells within 3D Engineered Matrices.
Tissue engineering. Part C, Methods (2015-01-17)
Bin Duan, Laura A Hockaday, Shoshana Das, Charlie Xu, Jonathan T Butcher
ABSTRACT
Living tissue-engineered heart valves (TEHV) would be a major benefit for children who require a replacement with the capacity for growth and biological integration. A persistent challenge for TEHV is accessible human cell source(s) that can mimic native valve cell phenotypes and matrix remodeling characteristics that are essential for long-term function. Mesenchymal stem cells derived from bone marrow (BMMSC) or adipose tissue (ADMSC) are intriguing cell sources for TEHV, but they have not been compared with pediatric human aortic valve interstitial cells (pHAVIC) in relevant 3D environments. In this study, we compared the spontaneous and induced multipotency of ADMSC and BMMSC with that of pHAVIC using different induction media within three-dimensional (3D) bioactive hybrid hydrogels with material modulus comparable to that of aortic heart valve leaflets. pHAVIC possessed some multi-lineage differentiation capacity in response to induction media, but limited to the earliest stages and much less potent than either ADMSC or BMMSC. ADMSC expressed cell phenotype markers more similar to pHAVIC when conditioned in basic fibroblast growth factor (bFGF) containing HAVIC growth medium, while BMMSC generally expressed similar extracellular matrix remodeling characteristics to pHAVIC. Finally, we covalently attached bFGF to PEG monoacrylate linkers and further covalently immobilized in the 3D hybrid hydrogels. Immobilized bFGF upregulated vimentin expression and promoted the fibroblastic differentiation of pHAVIC, ADMSC, and BMMSC. These findings suggest that stem cells retain a heightened capacity for osteogenic differentiation in 3D culture, but can be shifted toward fibroblast differentiation through matrix tethering of bFGF. Such a strategy is likely important for utilizing stem cell sources in heart valve tissue engineering applications.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 1.8 gal (6.8 L)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 3.6 gal (13.6 L)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00