Sigma-Aldrich
Trifluoroacetic anhydride, ReagentPlus®, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
1,4-Diaminobutane, 99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Cadaverine, 95%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, natural, ≥97%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing