MilliporeSigma
  • Human microvascular endothelial cells displaying reduced angiogenesis and increased uptake of lipids during in vitro culture.
Human microvascular endothelial cells displaying reduced angiogenesis and increased uptake of lipids during in vitro culture.
Clinical hemorheology and microcirculation (2015-10-08)
Henrieke Sievers, Ruth M Hirschberg, Bernhard Hiebl, Hana Hünigen, Johanna Plendl
ABSTRACT
Human microvascular ECs from the neonatal foreskin of two donors purchased from one distributor were used in an angiogenesis assay under the same culture conditions. Different angiogenic potency was apparent in these two batches (ECang and ECnon-ang). During the cultivation period of three weeks, ECang ran through all stages of angiogenesis starting from proliferation to migration up to the formation of three-dimensional capillary-like structures. Despite of expression of endothelial markers, ECnon-ang showed excessive intracellular storage of lipids in form of multilamellar bodies and decreased angiogenic potency in contrast to its counterpart, ECang. Results indicate that lipid metabolism differs in ECang versus ECnon-ang. This study points up that these differences are based on the different donors and presents a novel and valuable model for the study of mechanisms of atherosclerosis in endothelial cells in vitro.
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, suitable for chromatography, ≥99.8%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Acetone, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Acetone, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Acetone, suitable for residue analysis, ≥99.5%
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%