MilliporeSigma
  • Development and Fit-for-Purpose Validation of a Soluble Human Programmed Death-1 Protein Assay.
Development and Fit-for-Purpose Validation of a Soluble Human Programmed Death-1 Protein Assay.
The AAPS journal (2015-05-01)
Yan G Ni, Xiling Yuan, John A Newitt, Jon E Peterson, Carol R Gleason, Jonathan Haulenbeek, Rasa Santockyte, Virginie Lafont, Frank Marsilio, Robert J Neely, Binodh DeSilva, Steven P Piccoli
ABSTRACT
Programmed death-1 (PD-1) protein is a co-inhibitory receptor which negatively regulates immune cell activation and permits tumors to evade normal immune defense. Anti-PD-1 antibodies have been shown to restore immune cell activation and effector function-an exciting breakthrough in cancer immunotherapy. Recent reports have documented a soluble form of PD-1 (sPD-1) in the circulation of normal and disease state individuals. A clinical assay to quantify sPD-1 would contribute to the understanding of sPD-1-function and facilitate the development of anti-PD-1 drugs. Here, we report the development and validation of a sPD-1 protein assay. The assay validation followed the framework for full validation of a biotherapeutic pharmacokinetic assay. A purified recombinant human PD-1 protein was characterized extensively and was identified as the assay reference material which mimics the endogenous analyte in structure and function. The lower limit of quantitation (LLOQ) was determined to be 100 pg/mL, with a dynamic range spanning three logs to 10,000 pg/mL. The intra- and inter-assay imprecision were ≤15%, and the assay bias (percent deviation) was ≤10%. Potential matrix effects were investigated in sera from both normal healthy volunteers and selected cancer patients. Bulk-prepared frozen standards and pre-coated Streptavidin plates were used in the assay to ensure consistency in assay performance over time. This assay appears to specifically measure total sPD-1 protein since the human anti-PD-1 antibody, nivolumab, and the endogenous ligands of PD-1 protein, PDL-1 and PDL-2, do not interfere with the assay.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Sigma-Aldrich
Ruthenium black
Pricing and availability is not currently available.
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Ruthenium on carbon, extent of labeling: 5 wt. % loading
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetophenone, ≥98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Pricing and availability is not currently available.
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Pricing and availability is not currently available.