MilliporeSigma
Preparation and characterisation of hexamidine salts.
International journal of pharmaceutics (2015-08-04)
Nicola Parisi, Paul J Matts, Rebecca Lever, Jonathan Hadgraft, Majella E Lane
ABSTRACT
Hexamidine diisethionate (HEX D) has been used in the personal care industry and in a number of over-the-counter (OTC) drug products as an antimicrobial agent since the 1950's. Recently, the compound has also been investigated for its beneficial effects on skin health. Surprisingly, there is only limited information describing the physicochemical properties of this compound in the literature. The objective of this work was therefore to conduct a comprehensive programme of characterisation of HEX D as well as its dihydrochloride salt (HEX H). HEX H was prepared from HEX D by a simple acid addition reaction. Both salts were characterised using Nuclear Magnetic Resonance (NMR), Differential scanning calorimetry (DSC), and Thermogravimetric analysis (TGA). A new high performance liquid chromatographic method was developed and validated for both compounds. The pH in aqueous solution as well as respective distribution coefficients between octanol and pH 7.4 buffer were also determined. Finally, solubility and short term stability studies were conducted in a range of solvents. NMR analysis confirmed the preparation of HEX H from HEX D. Thermal analysis indicated the melting points of HEX D and HEX H were 225°C and 266°C respectively. HPLC analysis confirmed the purity of both salts. LogD values at pH 7.4 were -0.74 for HEX D and -0.70 for HEX H respectively. The physicochemical properties of two HEX salts have been established using a range of analytical approaches. Detailed solubility and stability data have also been collated. This information will be useful in the design of novel formulations for targeted delivery of these compounds to the skin.
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Isopropyl myristate, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Octanol, SAJ first grade, ≥75.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 12 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 2 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Octanol, JIS special grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 6 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Isopropyl myristate, ≥90% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D