MilliporeSigma
  • The secretion patterns and roles of cardiac and circulating arginine vasopressin during the development of heart failure.
The secretion patterns and roles of cardiac and circulating arginine vasopressin during the development of heart failure.
Neuropeptides (2015-04-01)
Xuanlan Chen, Guihua Lu, Kaiyu Tang, Qinglang Li, Xiuren Gao
ABSTRACT
The aim of this study is to investigate local cardiac and circulating AVP secretion during heart failure and to determine whether AVP mediates ventricular remodeling. We assessed cardiac function and AVP levels of post-myocardial infarction (MI) heart-failure rats 3 weeks (n = 10), 4 weeks (n = 10), 6 weeks (n = 10), 9 weeks (n = 15) after the proximal left anterior descending coronary artery (LAD) ligation. Ten sham-operated rats were used as the control group. In vitro, cardiac microvascular endothelial cells (CMECs) were initiated from isolated Wistar rat hearts and subjected to Ang II to induce AVP expression and secretion. Besides, the effects of AVP stimulation on CMECs and cardiac fibroblasts (CFs) were studied using methylthiazol tetrazolium assay, Western blotting and real-time PCR. With cardiac dysfunction, plasma and local cardiac AVP, aldosterone levels increased over time, peaking at 9 weeks post-MI. AVP levels were negatively correlated with serum Na(+) and LVEF but positively correlated with LVEDD and myocardial hydroxyproline. In CMECs treated with Ang II, AVP mRNA and protein expression increased. In addition, AVP promoted CFs proliferation and up-regulated the expression of endothelin-1 and connective tissue growth factor. CMECs are the cellular sources of elevated local heart AVP stimulated with Ang II/AT1. An intrinsic cardiac AVP system exists. Local cardiac and circulating AVP secretion were enhanced by deteriorating cardiac function. AVP may promote ventricular remodeling. Thus, AVP could be an important mediator of myocardial fibrosis in late-stage heart failure.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
SR 49059, ≥97% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
N-Bromosuccinimide, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
[Arg8]-Vasopressin acetate salt, ≥95% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%