MilliporeSigma
  • Cardioprotective effects of early and late aerobic exercise training in experimental pulmonary arterial hypertension.
Cardioprotective effects of early and late aerobic exercise training in experimental pulmonary arterial hypertension.
Basic research in cardiology (2015-10-16)
Daniel Moreira-Gonçalves, Rita Ferreira, Hélder Fonseca, Ana Isabel Padrão, Nuno Moreno, Ana Filipa Silva, Francisco Vasques-Nóvoa, Nádia Gonçalves, Sara Vieira, Mário Santos, Francisco Amado, José Alberto Duarte, Adelino F Leite-Moreira, Tiago Henriques-Coelho
ABSTRACT
Clinical studies suggest that aerobic exercise can exert beneficial effects in pulmonary arterial hypertension (PAH), but the underlying mechanisms are largely unknown. We compared the impact of early or late aerobic exercise training on right ventricular function, remodeling and survival in experimental PAH. Male Wistar rats were submitted to normal cage activity (SED), exercise training in early (EarlyEX) and in late stage (LateEX) of PAH induced by monocrotaline (MCT, 60 mg/kg). Both exercise interventions resulted in improved cardiac function despite persistent right pressure-overload, increased exercise tolerance and survival, with greater benefits in EarlyEX+MCT. This was accompanied by improvements in the markers of cardiac remodeling (SERCA2a), neurohumoral activation (lower endothelin-1, brain natriuretic peptide and preserved vascular endothelial growth factor mRNA), metabolism and mitochondrial oxidative stress in both exercise interventions. EarlyEX+MCT provided additional improvements in fibrosis, tumor necrosis factor-alpha/interleukin-10 and brain natriuretic peptide mRNA, and beta/alpha myosin heavy chain protein expression. The present study demonstrates important cardioprotective effects of aerobic exercise in experimental PAH, with greater benefits obtained when exercise training is initiated at an early stage of the disease.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Nitro-L-tyrosine, crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium trichloroacetate, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(+)-Biotin 4-nitrophenyl ester, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, SAJ first grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, JIS special grade