MilliporeSigma
Metabolic signatures of renal cell carcinoma.
Biochemical and biophysical research communications (2015-04-04)
Hwee Ying Lim, Yin Mun Yip, Edmund Chiong, Ho Yee Tiong, Barry Halliwell, Kesavan Esuvaranathan, Kim Ping Wong
ABSTRACT
Clear cell renal cell carcinoma (ccRCC) is characterized by the constitutive up-regulation of the hypoxia inducible factor-1. One of its target enzymes, pyruvate dehydrogenase (PDH) kinase 1 (PDHK1) showed increased protein expression in tumor as compared to patient-matched normal tissues. PDHK1 phosphorylated and inhibited PDH whose enzymatic activity was severely diminished, depriving the TCA cycle of acetylCoA. We and others have shown a decrease in the protein expressions of all respiratory complexes alluding to a compromise in oxidative phosphorylation (OXPHOS). On the contrary, we found that key parameters of OXPHOS, namely ATP biosynthesis and membrane potential were consistently measurable in mitochondria isolated from ccRCC tumor tissues. Interestingly, an endogenous mitochondrial membrane potential (MMP) was evident when ADP was added to mitochondria isolated from ccRCC but not in normal tissues. In addition, the MMP elicited in the presence of ADP by respiratory substrates namely malate/glutamate, succinate, α-ketoglutarate and isocitrate was invariably higher in ccRCC. Two additional hallmarks of ccRCC include a loss of uncoupling protein (UCP)-2 and an increase in UCP-3. Based on our data, we proposed that inhibition of UCP3 by ADP could contribute to the endogenous MMP observed in ccRCC and other cancer cells.
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)