MilliporeSigma
  • Signal transducers and activators of transcription 3-induced metastatic potential in gastric cancer cells is enhanced by glycogen synthase kinase-3β.
Signal transducers and activators of transcription 3-induced metastatic potential in gastric cancer cells is enhanced by glycogen synthase kinase-3β.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2015-04-08)
Jiyeon Yoon, Young San Ko, Sung Jin Cho, Jinju Park, Young Sun Choi, Yiseul Choi, Jung-Soo Pyo, Sang-Kyu Ye, Hong-Duk Youn, Jae-Seon Lee, Mee Soo Chang, Min A Kim, Byung Lan Lee
ABSTRACT
The transcription factor signal transducers and activators of transcription 3 (STAT3) can promote cancer metastasis, but its underlying regulatory mechanisms in gastric cancer cell invasiveness still remain obscure. We investigated the relationship between STAT3 and glycogen synthase kinase-3β (GSK-3β) and its significance in metastatic potential in gastric cancer cells. Immunohistochemical tissue array analysis of 267 human gastric carcinoma specimens showed that the expressions of active forms of STAT3 (pSTAT3) and GSK-3β (pGSK-3β) were found in 68 (25%) and 124 (46%) of 267 gastric cancer cases, respectively, showing a positive correlation (p < 0.001). Cell culture experiments using gastric cancer cell lines SNU-638 and SNU-668 revealed that STAT3 suppression did not affect pGSK-3β expression, whereas GSK-3β inhibition reduced pSTAT3 expression. With respect to metastatic potential in gastric cancer cells, both STAT3 suppression and GSK-3β inhibition decreased cell migration, invasion, and mesenchymal marker (Snail, Vimentin, and MMP9) expression. Moreover, the inhibitory effects of STAT3 and GSK-3β on cell migration were synergistic. These results demonstrated that STAT3 and GSK-3β are positively associated and synergistically contribute to metastatic potential in gastric cancer cells. Thus, dual use of STAT3 and GSK-3β inhibitors may enhance the efficacy of the anti-metastatic treatment of gastric cancer.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
AR-A014418, ≥98% (HPLC), solid
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stat3
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Bromophenol Blue, JIS special grade
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC