MilliporeSigma
Interleukin 6 inhibits HBV entry through NTCP down regulation.
Virology (2015-03-15)
Fidaa Bouezzedine, Olivier Fardel, Philippe Gripon
ABSTRACT
Hepatitis B virus (HBV) infection is a major public health problem. Recently, the human liver bile acid transporter Na(+)/taurocholate cotransporting polypeptide (NTCP) has been identified as an HBV specific receptor. NTCP expression is known to be strongly regulated by IL-6. This study was aimed at characterizing the effect of IL-6 on HBV entry. HBV entry was inhibited by up to 90% when cells were pretreated with IL-6 as shown by a strong inhibition of long term HBsAg secretion. This effect was confirmed by showing a severe reduction of intracellular HBV cccDNA. In parallel, we observed a 98% decrease in NTCP mRNA steady state level and an 80% reduction in NTCP-mediated taurocholate uptake. IL-6-mediated inhibition of NTCP-mediated taurocholate uptake and viral entry exhibited similar dose-dependence and kinetics while restoration of NTCP expression suppressed the inhibitory effect of IL-6. NTCP-mediated HBV entry is therefore markedly inhibited by IL-6.
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%