MilliporeSigma
Toxicity of Gossypol from Cottonseed Cake to Sheep Ovarian Follicles.
PloS one (2015-11-26)
Antônio Carlos Lopes Câmara, Ivana Cristina Nunes Gadelha, Pedro Augusto Cordeiro Borges, Silvano Alves de Paiva, Marília Martins Melo, Benito Soto-Blanco
ABSTRACT
Gossypol, a polyphenol compound produced by cotton plant, has proven reproductive toxicity, but the effects of gossypol on sheep ovaries are unknown. This study was aimed to determine the in vitro and in vivo effects of gossypol on the ovarian follicles of sheep. This trial was divided into two experiments. In the first one, we used twelve non-pregnant, nulliparous, Santa Inês crossbred ewes, which were randomly distributed into two equal groups and fed diets with and without cottonseed cake. Feed was offered at 1.5% of the animal's body weight for 63 days. The concentrations of total and free gossypol in the cottonseed cake were 3.28 mg/g and 0.11 mg/g, respectively. Throughout the trial period, no animal showed clinical signs of toxicity and no effects on body weight were observed. However, there was a significantly lower number of viable ovarian follicles (20.6%) and higher number of atretic follicles (79.4%) in the gossypol-fed sheep compared to the control (85.1 and 34.9%, respectively). These findings were observed at all stages of follicular development. In the second experiment, eight ovaries from slaughterhouse were cultured with different concentrations of gossypol acetic acid (0, 5, 10 and 20 μg/mL) for 24 hours or seven days. The in vitro action of gossypol resulted in a significant decrease in viable ovarian follicles, especially the primary and transition follicles, and a significant increase in the number of atretic follicles after 24 hours of culture. These follicles were greatly affected when cultured with gossypol for seven days. It is concluded that gossypol present in cotton seeds directly acts on ovarian follicles in sheep to increase atresia.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, FCC, FG