MilliporeSigma
HomeCell Culture TroubleshootingCell Culture Antibiotic Selection Guide
Cell Culture Antibiotic Selection Guide
Antibiotics are commonly used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications. The use of antibiotics can minimize loss of cell lines and cell cultures, saving time and conserving precious resources. Proper antibiotic selection depends on both cell type and which contaminants you are trying to prevent. Selective antibiotics for generating stable cell lines or other recombinant cultures should be chosen based on the antibiotic resistance gene or selectable marker. The tables below can be used to guide antibiotic selection for your cell culture needs.
Antibiotic by Target
Cell Wall Synthesis
Inhibit
Classification
Product Type
Product No.
Antibiotic Spectrum
Gram -ve
beta-lactamase
Penicillins
Penicillin
P3032
x
Penicillins
Penicillin
P7794
x
peptidoglycan formation
Glycopeptides
Vancomycin
V1130
x
Inhibit
Classification
Product Type
Product No.
Antibiotic Spectrum
Gram -ve
Fungi
Yeast
membrane phospolipids
Polymixins
Polymixin B
P1189
x
P4932
x
ergosterol biosynthesis
Polyenes
Amphotericin B
A9528
x
x
A2411
x
x
A2942
x
x
E3789
x
x
V900919
x
x
Inhibit
Classification
Product Type
Product  No.
Antibiotic Spectrum
Selection Antibiotic
Gram -ve
Gram +ve
Fungi
Mycoplasma
30S ribosome subunit
Tetracyclines
Tetracycline
T7660
x
x
x
Doxycycline
D5207
x
x
x
Aminocyclitol
Spectinomycin
S4014
x
x
x
S6258
x
x
x
Hygromycin
H3274
x
x
x
H9773
x
x
x
polypeptide synthesis
Aminoglycosides
Streptomycin
S1277
x
x
D5155
x
x
Paromomycin
P5057
x
x
P8692
x
x
Gentamicin
G1264
x
x
x
G1272
x
x
x
G1397
x
x
x
G1522
x
x
x
G9654
x
x
x
Kanamycin
K0254
x
x
x
x
K0129
x
x
x
x
Neomycin
N1142
x
x
x
N3144
x
x
x
N6386
x
x
x
G418
A1720
x
G1279
x
G8168
x
Nourseothricin
N0186
x
x
x
x
50S ribosome subunit
Macrolides
Tylosin
T6271
x
x
T3397
x
x
Erythromycin
E5389
x
Nystatin
N1638
x
N4014
x
N6261
x
N6803
x
N9150
x
Lincomycin
L2774
x
Rifampicin
R7382
x
x
peptidyl transferase
Chloramphenicols
Chloramphenicol
C3175
x
x
x
x
Inhibit
Classification
Product Type
Product No.
Antibiotic Spectrum
Selection Antibiotic
Gram -ve
Gram +ve
Cancer cells
DNA synthesis
IMPDH inhibitors
Mycophenolic acid
M3536
x
PKC inhibitors
Tamoxifen
T2859
x
ssDNA scisson
Glycopeptides
Bleomycin
B7216
x
Glycopeptides
Bleomycin
B8416
x
RNA synthesis
Amino-nucleoside
Puromycin
P8833
x
x
x
Amino-nucleoside
Puromycin
P4512
x
x
x
Aziridines
Mitomycin C
M4287
x
x
x
DNA structure through alkylation
Aziridines
Mitomycin C
M5353
x
x
x
Anthracyclines
Doxorubicin
D5220
x
purine biosynthesis
DHFR inhibitors
Methotrexate
M8407
x
Inhibit
Classification
Product Type
Product No.
Antibiotic Spectrum
Gram -ve
Gram +ve
beta-lactamase & polypeptide synthesis
Penicillins & aminoglycosides
Penicillin-Streptomycin
G1146
x
x
G6784
x
x
P0781
x
x
P4333
x
x
P4458
x
x
P7539
x
x
TMS-AB2-C
x
x
V900929
x
x
Inhibit
Classification
Product Type
Product No.
Antibiotic Spectrum
Gram -ve
Gram +ve
Fungi
Yeast
beta-lactamase, membrane phospholipids & polypeptide synthesis
Penicillins & polyenes
Antibiotic-Antimycotic solution
A5955
x
x
x
x