Skip to Content
Merck
All Photos(1)

Key Documents

A9219

Sigma-Aldrich

Agitoxin-2 from scorpion venom

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

assay

≥98% (SDS-PAGE)

form

lyophilized powder

mol wt

4015 Da

Storage temp.

−20°C

Biochem/physiol Actions

Potent Shaker K+ channel blocker . Useful as a probe for Kv channels.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Reconstitution

Reconstitute with 10 mM Tris, pH 7.5, containing 100 mM sodium chloride, 0.1% bovine serum albumin, and 1 mM EDTA.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Garcia et al.
Biochemistry, 33(22), 6834-6839 (1994-06-07)
Three new toxins from the venom of the scorpion Leiurus quinquestriatus var. hebraeus have been identified on the basis of their ability to block the Shaker K+ channel. These toxins have been purified using HPLC techniques and characterized as 38
Starlee Lively et al.
Frontiers in cellular neuroscience, 12, 115-115 (2018-05-22)
The cytokine, transforming growth factor β1 (TGFβ1), is up-regulated after central nervous system (CNS) injuries or diseases involving microglial activation, and it has been proposed as a therapeutic agent for treating neuroinflammation. Microglia can produce and respond to TGFβ1. While

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service