Skip to Content
Merck
All Photos(1)

Key Documents

SRP4589

Sigma-Aldrich

FGF-20 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

FGF20, FGFK, Fibroblast growth factor 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

lyophilized

mol wt

~24 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF20(26281)

General description

The gene FGF20 (fibroblast growth factor 20) is mapped to human chromosome 8p22-p21.3. It is strongly expressed in the substantia nigra of the brain. It is also expressed during early embryonic development. FGF20 belongs to the FGF class of genes.

Biochem/physiol Actions

FGF20 (fibroblast growth factor 20) is a neurotrophin which plays an important role in growth and survival of neurons. It is mainly required for the survival of dopaminergic cells. It plays an important role in central nervous development and function. FGF20 interacts with the tyrosine kinase FGF receptor-1 (FGFR1). Mutation in FGF20 (rs12720208) is associated with the susceptibility of Parkinson′s disease. It is also a putative proto-oncogene. It is required for maintaining the Wnt/β-catenin pathway-mediated anchorage-independent cellular growth.

Physical form

Lyophilized from 20 mM Tris, pH 7.6, containing 1.5 M NaCl.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1 to 1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fibroblast growth factor 20 polymorphisms and haplotypes strongly influence risk of Parkinson disease.
van der Walt JM, et al.
American Journal of Human Genetics, 74, 1121-1121 (2004)
Variation in the miRNA-433 binding site of FGF20 confers risk for Parkinson disease by overexpression of alpha-synuclein.
Wang G, et al.
American Journal of Human Genetics, 82, 283-283 (2008)
FGF-20 and DKK1 are transcriptional targets of beta-catenin and FGF-20 is implicated in cancer and development.
Chamorro MN, et al.
The Embo Journal, 24, 73-73 (2005)
Quantitative assessment of the effect of FGF20 rs12720208 variant on the risk of Parkinson's disease: a meta-analysis.
Wang X, et al.
Neurological Research, 39, 374-374 (2017)
Genetic variation in FGF20 modulates hippocampal biology.
Lemaitre H, et al.
The Journal of Neuroscience, 30, 5992-5992 (2010)

Articles

Fibroblast growth factors in cell culture and various growth factors for your research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service