MilliporeSigma
  • Effect of Lonicerae Flos extracts on reflux esophagitis with antioxidant activity.
Effect of Lonicerae Flos extracts on reflux esophagitis with antioxidant activity.
World journal of gastroenterology (2009-10-14)
Sae-Kwang Ku, Bu-Il Seo, Ji-Ha Park, Gyu-Yeol Park, Young-Bae Seo, Jae-Soo Kim, Hyeung-Sik Lee, Seong-Soo Roh
ABSTRACT
To observe the effects of traditional antiinflammatory medicine Lonicerae Flos (LF) on rat reflux esophagitis (RE) induced by pylorus and forestomach ligation compared with the well-known proton antioxidant, alpha-tocopherol. Rats were pretreated with three different dosages of LF (500, 250 and 125 mg/kg) orally, once a day for 14 d before pylorus and forestomach ligation. Nine hours after pylorus and forestomach ligation, changes to the stomach and esophagus lesion areas, gastric volumes, acid and pepsin outputs, antioxidant effects, esophageal lipid peroxidation, superoxide dismutase (SOD), catalase (CAT), malondialdehyde (MDA), myeloperoxidase and glutathione (GSH) levels, and collagen contents (marker of flexibility) were observed on the esophageal and fundic histopathology. The results were compared with an alpha-tocopherol (once orally, 1 h before operation, 30 mg/kg) treated group in which the effects on RE were already confirmed. Pylorus and forestomach ligations caused marked increases of gross esophageal and gastric mucosa lesion areas, which corresponded with histopathological changes. In addition, increases of esophageal lipid peroxidation, decreases of SOD, CAT, and GSH-free radical scavengers, increases of collagen were observed. However, these pylorus and forestomach ligation induced RE were dose-dependently inhibited by treatment of 500, 250 and 125 mg/kg of LF extract, mediated by antioxidant effects. RE at 250 mg/kg showed similar effects alpha-tocopherol. The results suggest that antioxidant effects of LF could attenuate the severity of RE and prevent the esophageal mucosal damage, and validate its therapeutic use in esophageal reflux disease.
Product Description
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyrogallol, JIS special grade, ≥99.0%
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Supelco
Pyrogallol, analytical standard
Sigma-Aldrich
Corn oil, CP
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Supelco
Corn oil, analytical standard
Supelco
Fast Green FCF, analytical standard
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
Fast Green FCF, Dye content ≥85 %
Sigma-Aldrich
Fast Green FCF, certified by the Biological Stain Commission
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds