MilliporeSigma
  • The Role of auxin, pH, and stress in the activation of embryogenic cell division in leaf protoplast-derived cells of alfalfa.
The Role of auxin, pH, and stress in the activation of embryogenic cell division in leaf protoplast-derived cells of alfalfa.
Plant physiology (2002-08-15)
Taras P Pasternak, Els Prinsen, Ferhan Ayaydin, Pál Miskolczi, Geert Potters, Han Asard, Harry A Van Onckelen, Dénes Dudits, Attila Fehér
ABSTRACT
Culturing leaf protoplast-derived cells of the embryogenic alfalfa (Medicago sativa subsp. varia A2) genotype in the presence of low (1 microM) or high (10 microM) 2, 4-dichlorophenoxyacetic acid (2,4-D) concentrations results in different cell types. Cells exposed to high 2,4-D concentration remain small with dense cytoplasm and can develop into proembryogenic cell clusters, whereas protoplasts cultured at low auxin concentration elongate and subsequently die or form undifferentiated cell colonies. Fe stress applied at nonlethal concentrations (1 mM) in the presence of 1 microM 2,4-D also resulted in the development of the embryogenic cell type. Although cytoplasmic alkalinization was detected during cell activation of both types, embryogenic cells could be characterized by earlier cell division, a more alkalic vacuolar pH, and nonfunctional chloroplasts as compared with the elongated, nonembryogenic cells. Buffering of the 10 microM 2,4-D-containing culture medium by 10 mM 2-(N-morpholino)ethanesulfonic acid delayed cell division and resulted in nonembryogenic cell-type formation. The level of endogenous indoleacetic acid (IAA) increased transiently in all protoplast cultures during the first 4 to 5 d, but an earlier peak of IAA accumulation correlated with the earlier activation of the division cycle in embryogenic-type cells. However, this IAA peak could also be delayed by buffering of the medium pH by 2-(N-morpholino)ethanesulfonic acid. Based on the above data, we propose the involvement of stress responses, endogenous auxin synthesis, and the establishment of cellular pH gradients in the formation of the embryogenic cell type.
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
MES sodium salt, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
MES sodium salt, BioReagent, ≥99% (titration), crystalline
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
MES sodium salt, ≥99% (titration)
SAFC
MES sodium salt
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)