MilliporeSigma
  • Effect of heat moisture treatment on physicochemical and morphological properties of wheat starch and xylitol mixture.
Effect of heat moisture treatment on physicochemical and morphological properties of wheat starch and xylitol mixture.
Food chemistry (2013-09-24)
Qingjie Sun, Lei Dai, Chong Nan, Liu Xiong
ABSTRACT
The effect of heat moisture treatment (HMT, 100°C, 10h) on the physicochemical and morphological properties of wheat starch and xylitol mixture (SX) were studied by using Rapid Visco-Analyser (RVA), differential scanning calorimetry (DSC), scanning electron microscope (SEM) and X-ray diffraction pattern (X-ray). The research found that the peak viscosity (PV), trough viscosity (TV), final viscosity (FV) and breakdown (BD) of SX decreased markedly after HMT, whereas pasting temperature increased. Compared with SX, To, Tp and Tc of SX treated with HMT (H-SX) increased and ΔH decreased. The results of SEM showed that numerous xylitol granules appeared on the surface of starch granule in SX, and a layer of similar membrane adhered on the surface of starch granules in H-SX. After gelatinization and freeze-dried, the gel network structure of H-SX was tighter than that of SX. The amorphous region of H-SX was higher than that of SX.
Sigma-Aldrich
Starch from corn, practical grade
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Xylitol, ≥99% (GC)
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Xylitol
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material