MilliporeSigma
  • Cytotoxic T lymphocyte-associated antigen 4 inhibition increases the antitumor activity of adoptive T-cell therapy when carried out with naïve rather than differentiated T cells.
Cytotoxic T lymphocyte-associated antigen 4 inhibition increases the antitumor activity of adoptive T-cell therapy when carried out with naïve rather than differentiated T cells.
Oncology reports (2015-03-05)
Takeshi Ishikawa, Satoko Adachi, Tetsuya Okayama, Satoshi Kokura, Katsura Mizushima, Toshifumi Doi, Tatsuzo Matsuyama, Naoyuki Sakamoto, Kazuhiro Katada, Kazuhiro Kamada, Kazuhiko Uchiyama, Osamu Handa, Tomohisa Takagi, Yuji Naito, Yoshito Itoh, Toshikazu Yoshikawa
ABSTRACT
Although treatment with an antibody against cytotoxic T-lymphocyte-associated antigen 4 (CTLA-4) combined with multiple therapeutic interventions has been explored, the effect of combination therapy with CTLA-4 inhibition and adoptive T-cell therapy has not been determined. In the present study, our aim was to determine whether CTLA-4 inhibition, combined with adoptive transfer of T cells at different stages of differentiation, exhibits synergistic antitumor effects in a murine colon cancer model. Mice bearing subcutaneous tumors were administered adoptive T-cell transfer of CD62Lhigh or CD62Llow cells combined with an anti-CTLA-4 antibody (α-CTLA-4) or control immunoglobulin G. Subcutaneous tumors were harvested, and the antitumor effects and helper T-cell polarization were analyzed. CTLA-4 inhibition combined with CD62Lhigh cell administration showed the strongest antitumor effect. Combination therapy increased the number of CD3+ cells within the tumor. Moreover, CTLA-4 inhibition induced polarization of T cells infiltrating the tumor toward the T helper 1 lineage, and suppressed the frequency of regulatory T cells within the tumor, particularly in combination with CD62Lhigh T-cell transfer. This is the first report demonstrating that the efficacy of α-CTLA-4 and adoptive T-cell transfer combination therapy depends on the state of differentiation of the transferred T cells. Our data support the notion that a combination of α-CTLA-4 and adoptive T-cell transfer containing an abundance of naïve phenotype cells could potentially exert antitumor effects in a clinical setting.
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Methanol, NMR reference standard
SAFC
BIS-TRIS
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Interleukin-7 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citric acid trisodium salt, ≥98% (GC), anhydrous
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
BIS-TRIS
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free