MilliporeSigma
  • Labeled content of two furanocoumarins in dietary supplements correlates with neither actual content nor CYP3A inhibitory activity.
Labeled content of two furanocoumarins in dietary supplements correlates with neither actual content nor CYP3A inhibitory activity.
Journal of pharmaceutical and biomedical analysis (2014-06-22)
Karen M VanderMolen, Garrett R Ainslie, Mary F Paine, Nicholas H Oberlies
ABSTRACT
Dietary supplements are a multi-billion dollar business, with yearly profit increases. Allegedly safe, these supplements are marketed to a variety of niches, encompassing claims from immune support to weight loss. Six sports nutrition supplements were acquired that were labeled to contain the furanocoumarin(s) bergamottin and/or 6',7'-dihydroxybergamottin (DHB), both of which are potent irreversible inhibitors of the prominent drug metabolizing enzyme cytochrome P450 3A (CYP3A). Both furanocoumarins are typically present in grapefruit juice, which has been shown to inhibit intestinal CYP3A, perpetrating an increase in the systemic exposure of certain concomitant 'victim' drugs. The acquired supplements were analyzed using ultra-performance liquid chromatography coupled to both a photodiode array (PDA) detector and a triple quadrupole mass spectrometer (MS). Contrary to the product labeling, four of the supplements contained no detectable quantities of either furanocoumarin (LOD 0.060μg/capsule), while two of the supplements contained minimal amounts (one contained 12.13 (±0.23) μg bergamottin and 65.51 (±0.64) μg DHB per capsule; the other contained 2.705 (±0.069) μg bergamottin per capsule and no detectable quantities of DHB). A CYP3A inhibition bioassay was used to assess whether the actual content of the furanocoumarins correlated with CYP3A inhibitory activity. Despite the low amounts of bergamottin and DHB, CYP3A inhibition by the supplements was greater than could be accounted for by the two furanocoumarins. The additional activity suggests the presence of other potent or highly abundant CYP3A inhibitors.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Alprazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Alprazolam, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Alprazolam
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%