MilliporeSigma
  • APL-2, an altered peptide ligand derived from heat-shock protein 60, induces interleukin-10 in peripheral blood mononuclear cell derived from juvenile idiopathic arthritis patients and downregulates the inflammatory response in collagen-induced arthritis model.
APL-2, an altered peptide ligand derived from heat-shock protein 60, induces interleukin-10 in peripheral blood mononuclear cell derived from juvenile idiopathic arthritis patients and downregulates the inflammatory response in collagen-induced arthritis model.
Clinical and experimental medicine (2014-01-30)
Norailys Lorenzo, Dolores Cantera, Ariana Barberá, Amaris Alonso, Elsy Chall, Lourdes Franco, Julio Ancizar, Yanetsy Nuñez, Fiorella Altruda, Lorenzo Silengo, Gabriel Padrón, Maria Del Carmen Dominguez
ABSTRACT
Juvenile idiopathic arthritis (JIA) is a heterogeneous group of diseases characterized by autoimmune arthritis of unknown cause with onset before age of 16 years. Methotrexate provides clinical benefits in JIA. For children who do not respond to methotrexate, treatment with anti-tumor necrosis factor (TNF)-α is an option. However, some patients do not respond or are intolerant to anti-TNF therapy. Induction of peripheral tolerance has long been considered a promising approach to the treatment of chronic autoimmune diseases. We aimed to evaluate the potentialities of two altered peptide ligands (APLs) derived from human heat-shock protein 60, an autoantigen involved in the pathogenesis of autoimmune arthritis, in JIA patients. Interferon (IFN)-γ, TNF-α and interleukin (IL)-10 levels were determined in ex vivo assays using peripheral blood mononuclear cells (PBMC) from these patients. Wild-type peptide and one of these APLs increased IFN-γ and TNF-α levels. Unlike, the other APLs (called APL2) increased the IL-10 level without affecting IFN-γ and TNF-α levels. On the other hand, APL2 induces a marked activation of T cells since it transforms cell cycle phase's distribution of CD4+ T cells from these patients. In addition, we evaluated the therapeutic effect of APL2 in collagen-induced arthritis model. Therapy with APL2 reduced arthritis scores and histological lesions in mice. This effect was associated to a decrease in TNF-α and IL-17 levels. These results indicate a therapeutic potentiality of APL2 for JIA.
Sigma-Aldrich
Propidium iodide solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
SAFC
L-Glutamine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
L-Glutamine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
SAFC
HEPES
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
SAFC
HEPES
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 23, 2025
$207.00