MilliporeSigma
  • Antisense-mediated angiotensinogen inhibition slows polycystic kidney disease in mice with a targeted mutation in Pkd2.
Antisense-mediated angiotensinogen inhibition slows polycystic kidney disease in mice with a targeted mutation in Pkd2.
American journal of physiology. Renal physiology (2014-12-30)
Kameswaran Ravichandran, Abdullah Ozkok, Qian Wang, Adam E Mullick, Charles L Edelstein
ABSTRACT
Renal cyst enlargement is associated with the activation of both the circulating and intrarenal renin-angiotensin systems. Angiotensinogen (AGT) is the substrate for renin. The aim of the present study was to determine the effect of AGT inhibition on renal cyst enlargement. An AGT antisense oligonucleotide (ASO) that selectively inhibits AGT mRNA was injected once weekly in PKD2WS25 mice [an orthologous model of human autosmal dominant polycystic kidney disease (PKD) involving mutation of the Pkd2 gene] from 4 to 16 wk of age. The AGT ASO resulted in a 40% decrease in AGT RNA in the kidney, a 60% decrease in AGT RNA in the liver, and a significant decrease in AGT protein in the kidney and serum. The AGT ASO resulted in a significant decrease in kidney size, cyst volume density, and blood urea nitrogen. The AGT ASO resulted in a significant decrease in transforming growth factor-β and interstitial fibrosis in the kidney. Mice treated with the AGT ASO had a significant decrease in proinflammatory cytokines [chemokine (C-X-C motif) ligand (CXCL)1 and IL-12] in the kidney. Cluster of differentiation (CD)36 is a scavenger receptor found on tubular cells that can activate the renin-angiotensin system. Administration of a CD36 ASO had no effect on PKD and kidney function, suggesting that the effect of the AGT ASO is independent of CD36. In summary, AGT inhibition resulted in significant decreases in kidney size and cyst volume and an improvement in kidney function in PKD mice. The AGT ASO resulted in a decrease in transforming growth factor-β, interstitial fibrosis, and the proinflammatory cytokines CXCL1 and IL-12 in the kidney.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Picric acid solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Picric acid solution
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Millipore
Urea solution, suitable for microbiology, 40% in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Picric acid, moistened with water, ≥98%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture