MilliporeSigma
  • Rapid extraction and determination of amphetamines in human urine samples using dispersive liquid-liquid microextraction and solidification of floating organic drop followed by high performance liquid chromatography.
Rapid extraction and determination of amphetamines in human urine samples using dispersive liquid-liquid microextraction and solidification of floating organic drop followed by high performance liquid chromatography.
Journal of pharmaceutical and biomedical analysis (2014-03-04)
Toraj Ahmadi-Jouibari, Nazir Fattahi, Mojtaba Shamsipur
ABSTRACT
A novel, rapid, simple and sensitive dispersive liquid-liquid microextraction method based on the solidification of floating organic drop (DLLME-SFO) combined with high-performance liquid chromatography-ultraviolet detection (HPLC-UV) was used to determine amphetamine and methamphetamine in urine samples. The factors affecting the extraction efficiency of DLLME-SFO such as the kind and volume of the extraction and the disperser solvents, effect of concentration of K2CO3 and extraction time were investigated and the optimal extraction conditions were established. Under the optimum conditions (extraction solvent: 30.0μl 1-undecanol; disperser solvent: 300μl acetonitrile; buffer concentration: 2% (w/v) K2CO3 and extraction time: 1min), calibration curves are linear in the range of 10-3000μgl(-1) and limit of detections (LODs) are in the range of 2-8μgl(-1). The relative standard deviations (RSDs) for 100μgl(-1) of amphetamine and methamphetamine in diluted urine are in the range of 6.2-7.8% (n=7). The method was successfully applied for the determination of amphetamine and methamphetamine in the actual urine samples. The relative recoveries of urine samples spiked with amphetamine and methamphetamine are 87.8-113.2%. The obtained results show that DLLME-SFO combined with HPLC-UV is a fast and simple method for the determination of amphetamine and methamphetamine in urine.
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Undecanol, 97%
Pricing and availability is not currently available.
Supelco
Hexadecane, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Pricing and availability is not currently available.
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Pricing and availability is not currently available.
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
2-Dodecanol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Decanol, ≥98%, FCC, FG
Pricing and availability is not currently available.
Supelco
Acetonitrile, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
1-Decanol, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Decanol, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Pricing and availability is not currently available.
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Pricing and availability is not currently available.
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Pricing and availability is not currently available.