MilliporeSigma
  • AP2 suppresses osteoblast differentiation and mineralization through down-regulation of Frizzled-1.
AP2 suppresses osteoblast differentiation and mineralization through down-regulation of Frizzled-1.
The Biochemical journal (2014-11-05)
Shibing Yu, Laura M Yerges-Armstrong, Yanxia Chu, Joseph M Zmuda, Yingze Zhang
ABSTRACT
Transcription factor activating protein 2 (AP2) plays an important role in cellular differentiation. Although profound craniofacial and long bone developmental abnormalities have been observed in AP2-knockout mice, the molecular effects of AP2 on osteoblasts are poorly defined. We demonstrated that AP2 regulates the expression of human Frizzled 1 (FZD1), a co-receptor for the Wnt signalling pathway, in human osteoblast cell lines and primary bone marrow stromal cells (BMSCs). We also identified a putative AP2-binding site in the FZD1 proximal promoter in silico and characterized this binding element further in Saos2 in vitro by ChIP, electrophoretic mobility shift and promoter reporter assays. The transcriptional repression of the FZD1 promoter by AP2 was confirmed in normal human fetal osteoblasts (hFOB). Furthermore, overexpression of AP2 resulted in a significant reduction in both differentiation and mineralization of Saos2 cells. Knockdown of FZD1 expression before AP2 up-regulation diminished the AP2-dependent inhibition of Saos2 cell differentiation and mineralization. Similarly, overexpressing FZD1 before AP2 treatment in both Saos2 and BMSCs diminished the inhibitory effect of AP2 on osteoblast differentiation and mineralization. Taken together, these results demonstrate that AP2 is a negative regulator of osteoblast differentiation and mineralization, and its inhibitory effect may be mediated in part through down-regulation of FZD1 expression.
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard