MilliporeSigma
  • Genetic deletion of neuronal pentraxin 1 expression prevents brain injury in a neonatal mouse model of cerebral hypoxia-ischemia.
Genetic deletion of neuronal pentraxin 1 expression prevents brain injury in a neonatal mouse model of cerebral hypoxia-ischemia.
Neurobiology of disease (2015-01-03)
Shabarish Thatipamula, Md Al Rahim, Jiangyang Zhang, Mir Ahamed Hossain
ABSTRACT
Neonatal hypoxic-ischemic (HI) brain injury is a leading cause of mortality and morbidity in infants and children for which there is no promising therapy at present. Previously, we reported induction of neuronal pentraxin 1 (NP1), a novel neuronal protein of the long-pentraxin family, following HI injury in neonatal brain. Here, we report that genetic deletion of NP1 expression prevents HI injury in neonatal brain. Elevated expression of NP1 was observed in neurons, not in astrocytes, of the ipsilateral cortical layers (I-IV) and in the hippocampal CA1 and CA3 areas of WT brains following hypoxia-ischemia; brain areas that developed infarcts (at 24-48 h), showed significantly increased numbers of TUNEL-(+) cells and tissue loss (at 7 days). In contrast, NP1-KO mice showed no evidence of brain infarction and tissue loss after HI. The immunofluorescence staining of brain sections with mitochondrial protein COX IV and subcellular fractionation analysis showed increased accumulation of NP1 in mitochondria, pro-death protein Bax activation and NP1 co-localization with activated caspase-3 in WT, but not in the NP1-KO brains; corroborating NP1 interactions with the mitochondria-derived pro-death pathways. Disruption of NP1 translocation to mitochondria by NP1-siRNA in primary cortical cultures significantly reduced ischemic neuronal death. NP1 was immunoprecipitated with activated Bax [6A7] proteins; HI caused increased interactions of NP1 with Bax, thereby, facilitating Bax translocation to mitochondrial and neuronal death. To further delineate the specificity of NPs, we found that NP1 but not the NP2 induction is specifically involved in brain injury mechanisms and that knockdown of NP1 only results in neuroprotection. Furthermore, live in vivo T2-weighted magnetic resonance imaging (MRI) including fractional anisotropy (FA) mapping showed no sign of delayed brain injury or tissue loss in the NP1-KO mice as compared to the WT at different post-HI periods (4-24 weeks) examined; indicating a long-term neuroprotective efficacy of NP1 gene deletion. Collectively, our results demonstrate a novel mechanism of neuronal death and predict that inhibition of NP1 expression is a promising strategy to prevent hypoxic-ischemic injury in immature brain.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, tablet
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride solution, 5 M
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride solution, 1 M
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing