MilliporeSigma
  • Flatworm models in pharmacological research: the importance of compound stability testing.
Flatworm models in pharmacological research: the importance of compound stability testing.
Regulatory toxicology and pharmacology : RTP (2014-07-08)
Sofie Stalmans, Maxime Willems, Els Adriaens, Jean-Paul Remon, Matthias D'Hondt, Bart De Spiegeleer
ABSTRACT
Flatworms possess adult pluripotent stem cells, which make them extraordinary experimental model organisms to assess in vivo the undesirable effects of substances on stem cells. Currently, quality practices, implying evaluation of the stability of the test compound under the proposed experimental conditions, are uncommon in this research field. Nevertheless, performing a stability study during the rational design of in vivo assay protocols will result in more reliable assay results. To illustrate the influence of the stability of the test substance on the final experimental outcome, we performed a short-term International Conference on Harmonization (ICH)-based stability study of cyclophosphamide in the culture medium, to which a marine flatworm model Macrostomum lignano is exposed. Using a validated U(H)PLC method, it was demonstrated that the cyclophosphamide concentration in the culture medium at 20°C is lowered to 80% of the initial concentration after 21days. The multiwell plates, flatworms and diatoms, as well as light exposure, did not influence significantly the cyclophosphamide concentration in the medium. The results of the stability study have practical implications on the experimental set-up of the carcinogenicity assay like the frequency of medium renewal. This case study demonstrates the benefits of applying appropriate quality guidelines already during fundamental research increasing the credibility of the results.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cyclophosphamide monohydrate, analytical standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Sigma-Aldrich
Cyclophosphamide monohydrate, ISOPAC®
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Acetonitrile, analytical standard