MilliporeSigma
  • Equol Inhibits LPS-Induced Oxidative Stress and Enhances the Immune Response in Chicken HD11 Macrophages.
Equol Inhibits LPS-Induced Oxidative Stress and Enhances the Immune Response in Chicken HD11 Macrophages.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-05-23)
Zhongyong Gou, Shouqun Jiang, Chuntian Zheng, Zhimei Tian, Xiajing Lin
ABSTRACT
There has been increasing recent attention on the antioxidative capacity of equol. This study tested the effect of equol on oxidative stress induced by lipopolysaccharide (LPS) and regulation of immunity in chicken macrophages. Chicken HD11 macrophages were challenged with LPS (100 ng/mL) alone or with LPS (100 ng/mL) and (±)equol (10, 20, 40, 80, 160 μmol/L) together for 24h. Evaluated responses included the contents of malondialdehyde (MDA) and reduced glutathione (GSH), activities of total superoxide dismutase (T-SOD) and inducible nitric oxide synthase (iNOS), transcript abundance of superoxide dismutase 2 (SOD2), catalase (CAT), glutathione transferase (GST), Toll-like receptor 4 (TLR4), tumor necrosis factor alpha (TNFα) and interleukin-1 beta (IL-1β), and contents of the cytokines TNFα, IL-1β, interleukin-2 (IL-2) and interferon beta (IFNβ). Exposure to LPS induced oxidative stress as contents of MDA increased and GSH decreased in LPS-treated cells (P < 0.05) compared to those in control cells. Compared to LPS alone, co-treatment with equol (20 μmol/L, 40 μmol/L or 80 μmol/L) reduced contents of MDA and increased those of GSH (both P < 0.05). Activity of T-SOD increased (P < 0.05) in cells treated with the higher contentration of equol (80 μmol/L or 160 μmol/L), however, all concentrations (20 μmol/L to 160 μmol/L) increased activity of iNOS (P < 0.05). The highest concentration of equol (160 μmol/L) increased SOD2 and GST transcripts (P < 0.05). Equol treatment increased transcripts of TLR4, TNFα and IL-1β (P < 0.05). And there were similar changes in contents of IL-1β, IL-2, IFNβ and TNFα in the cells (P < 0.05). It concluded that equol can protect chicken HD11 macrophages from oxidative stress induced by LPS through reducing lipid peroxidation products and enhancing contents of antioxidants, and activities of relevant antioxidase enzymes; effects were also seen in gene expression related to the immune response and increased contents of cytokines. The optimal concentration of equol on antioxidation and immune enhancement in chicken macrophages was 40 μmol/L.
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, SAJ special grade, ≥99.0%
SAFC
HEPES
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES