MilliporeSigma
  • Dispersive liquid-liquid microextraction for the determination of new generation pesticides in soils by liquid chromatography and tandem mass spectrometry.
Dispersive liquid-liquid microextraction for the determination of new generation pesticides in soils by liquid chromatography and tandem mass spectrometry.
Journal of chromatography. A (2015-04-07)
Marta Pastor-Belda, Isabel Garrido, Natalia Campillo, Pilar Viñas, Pilar Hellín, Pilar Flores, José Fenoll
ABSTRACT
A sensitive method for the determination of five new generation pesticides (chlorantraniliprole, spirotetramat, spiromesifen, spirodiclofen and flubendiamide) in soil samples has been developed using liquid chromatography and tandem mass spectrometry (LC-MS(2)) with a triple-quadrupole in the multiple reaction monitoring mode. The target analytes are released from the solid matrix by single-phase extraction in acetonitrile (SLE), this organic phase being used as dispersant solvent in the dispersive liquid-liquid microextraction (DLLME) step. The different parameters affecting the extraction efficiency in DLLME were carefully studied, being 1.5mL of acetonitrile extract (disperser solvent), 125μL carbon tetrachloride (extraction solvent) and 10mL aqueous solution, the selected conditions. The enriched organic phase was evaporated, reconstituted with 50μL acetonitrile and injected into a liquid chromatograph with a mobile phase composed of acetonitrile and 0.1% (v/v) formic acid under gradient elution and a C8 stationary phase. Detection limits in the 0.0015-0.0090ngg(-1) range were obtained. Insecticide concentrations ranging from 0.03 to 197ngg(-1), depending on the compound, were found in the soil samples analysed. The recovery values obtained by SLE-DLLME-LC-ESI-MS(2) for three spiked soils at three concentration levels varied between 87 and 114%, with RSDs of between 5.5 and 14%.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles