Merck
  • Effects of microbial phytase on apparent and standardized total tract digestibility of calcium in calcium supplements fed to growing pigs.
Effects of microbial phytase on apparent and standardized total tract digestibility of calcium in calcium supplements fed to growing pigs.
Journal of animal science (2015-05-29)
J C González-Vega, C L Walk, H H Stein
ABSTRACT
An experiment was conducted to test the hypothesis that differences in the apparent total tract digestibility (ATTD) and standardized total tract digestibility (STTD) of Ca exist among Ca supplements and that inclusion of microbial phytase increases the ATTD and STTD of Ca. One hundred and four growing barrows (average initial BW of 17.73 ± 2.53 kg) were allotted to a randomized complete block design with 13 dietary treatments and 8 pigs per treatment. A basal diet containing corn, cornstarch, potato protein isolate, soybean oil, calcium carbonate, monosodium phosphate, vitamins, and minerals was formulated. Five additional diets were formulated by adding monocalcium phosphate (MCP), dicalcium phosphate (DCP), calcium carbonate, Lithothamnium calcareum Ca, or a high-Ca sugar beet co-product to the basal diet at the expense of cornstarch. Six additional diets that were similar to the previous 6 diets with the exception that they also contained 500 units per kilogram of microbial phytase were also formulated. A Ca-free diet was used to determine basal endogenous losses of Ca. Feces were collected using the marker-to-marker approach. Results indicated that regardless of inclusion of microbial phytase, MCP had the greatest (P < 0.05) ATTD and STTD of Ca. The ATTD and STTD of Ca in DCP were greater (P < 0.05) than in calcium carbonate, L. calcareumC a, or in the sugar beet co-product, but no differences were observed among the ATTD and STTD of Ca in calcium carbonate, L. calcareum Ca, or sugar beet co-product. Inclusion of microbial phytase increased (P < 0.05) the ATTD and STTD of Ca in the diets, but this was not the case in the Ca supplements. Regardless of inclusion of microbial phytase, the ATTD of P was greater ( P< 0.05) in pigs fed basal, MCP, or DCP diets than in pigs fed calcium carbonate, L. calcareum Ca, or the sugar beet co-product, but pigs fed calcium carbonate diets had greater ( P< 0.05) ATTD of P than pigs fed L. calcareumCa or the sugar beet co-product. Regardless of Ca source, inclusion of microbial phytase increased (P < 0.001) the ATTD of P. In conclusion, MCP has the greatest ATTD and STTD of Ca among the calcium supplements used in this experiment, followed by DCP. Basal, MCP, and DCP diets had greater ATTD of P than the other diets, and inclusion of microbial phytase increased the ATTD and STTD of Ca and the ATTD of P in the diets.
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cholecalciferol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Tocopherol, ≥95.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, JIS special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium carbonate, CP
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, SAJ first grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Retinyl acetate, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sucrose, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g