MilliporeSigma
  • Preserved muscle oxidative metabolic phenotype in newly diagnosed non-small cell lung cancer cachexia.
Preserved muscle oxidative metabolic phenotype in newly diagnosed non-small cell lung cancer cachexia.
Journal of cachexia, sarcopenia and muscle (2015-07-03)
Celine M Op den Kamp, Harry R Gosker, Suzanne Lagarde, Daniel Y Tan, Frank J Snepvangers, Anne-Marie C Dingemans, Ramon C J Langen, Annemie M W J Schols
ABSTRACT
Cachexia augments cancer-related mortality and has devastating effects on quality of life. Pre-clinical studies indicate that systemic inflammation-induced loss of muscle oxidative phenotype (OXPHEN) stimulates cancer-induced muscle wasting. The aim of the current proof of concept study is to validate the presence of muscle OXPHEN loss in newly diagnosed patients with lung cancer, especially in those with cachexia. Quadriceps muscle biopsies of comprehensively phenotyped pre-cachectic (n = 10) and cachectic (n = 16) patients with non-small cell lung cancer prior to treatment were compared with healthy age-matched controls (n = 22). OXPHEN was determined by assessing muscle fibre type distribution (immunohistochemistry), enzyme activity (spectrophotometry), and protein expression levels of mitochondrial complexes (western blot) as well as transcript levels of (regulatory) oxidative genes (quantitative real-time PCR). Additionally, muscle fibre cross-sectional area (immunohistochemistry) and systemic inflammation (multiplex analysis) were assessed. Muscle fibre cross-sectional area was smaller, and plasma levels of interleukin 6 were significantly higher in cachectic patients compared with non-cachectic patients and healthy controls. No differences in muscle fibre type distribution or oxidative and glycolytic enzyme activities were observed between the groups. Mitochondrial protein expression and gene expression levels of their regulators were also not different. Muscle OXPHEN is preserved in newly diagnosed non-small cell lung cancer and therefore not a primary trigger of cachexia in these patients.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)