MilliporeSigma
  • Metronidazole reduces the expression of cytochrome P450 enzymes in HepaRG cells and cryopreserved human hepatocytes.
Metronidazole reduces the expression of cytochrome P450 enzymes in HepaRG cells and cryopreserved human hepatocytes.
Xenobiotica; the fate of foreign compounds in biological systems (2014-12-04)
Toshiyuki Kudo, Yumiko Endo, Rina Taguchi, Masami Yatsu, Kiyomi Ito
ABSTRACT
1. Blood levels of S-warfarin have been reported to be increased by concomitant administration of metronidazole (MTZ), an antiprotozoal imidazole derivative. 2. To elucidate the mechanism of this interaction and to identify other possible drug-drug interactions, we conducted an in vitro study with the human hepatoma HepaRG cells and cryopreserved human hepatocytes on the ability of MTZ to reduce the expression of cytochrome P450 (CYP) as well as nuclear receptors that regulate the expression of these enzymes. 3. HepaRG cells and cryopreserved human hepatocytes were treated with MTZ (20 to 500 µM) and were then analyzed by real-time RT-PCR to determine mRNA levels of drug-metabolizing enzymes and nuclear receptors. 4. In both cells, the expressions of CYP2C8, CYP2C9, CYP3A4 and constitutive androstane receptor (CAR) were decreased by MTZ treatment. Particularly, in HepaRG cells, their mRNA levels were decreased by MTZ treatment in a concentration-dependent manner. 5. Our findings suggest that the interaction between MTZ and S-warfarin may be due to the MTZ-induced down-regulation of CYP2C9, the primary enzyme responsible for S-warfarin hydroxylation, and CAR, which regulates CYP2C9 expression. We also found that MTZ use may alter the disposition of drugs metabolized by the CYP isozymes investigated.
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Metronidazole, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Metronidazole, BioXtra
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri