Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sign Into View Organizational & Contract Pricing Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Nitrogen, ≥99.998%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile, for chromatography
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing