MilliporeSigma
  • Effects of high-protein or conventional canola meal on growth performance, organ weights, bone ash, and blood characteristics of weanling pigs.
Effects of high-protein or conventional canola meal on growth performance, organ weights, bone ash, and blood characteristics of weanling pigs.
Journal of animal science (2015-05-29)
C K Parr, Y Liu, C M Parsons, H H Stein
ABSTRACT
An experiment was conducted to evaluate effects of 2 high-protein canola meals (canola meal A [CMA]: 45.69% CP and canola meal B [CMB]: 46.97% CP) and a conventional canola meal (CM-CV: 35.10% CP) on growth performance, organ weights, bone ash, and blood parameters of weanling pigs. Inclusion rates of canola meal (CM) in the diets were 10, 20, 30, or 40% for CMA and CM-CV, whereas inclusions were 10, 20, or 30% for CMB. A control diet containing no CM was also formulated. Therefore, 12 diets were used in this experiment. A total of 420 pigs (initial BW: 9.8 ± 1.1 kg) were divided into 3 blocks and randomly allotted to 1 of the 12 diets with 8 replicate pens per treatment and 4 or 5 pigs per pen. The ADG, ADFI, and G:F were calculated, and at the conclusion of the experiment, 1 pig in each pen was euthanized to allow measurements of organ weights, collection of blood, and collection of the third and fourth metacarpals from the left foot. Results indicate that ADFI was linearly (P < 0.05) decreased if inclusion of CMA, CMB, or CM-CV was increased. Average daily gain for pigs fed CMA tended to increase quadratically with the maximum response observed at 10 or 20% CM inclusion in the diet ( P= 0.06). However, G:F was linearly (P < 0.05) increased by adding CMA or CM-CV to the diets. Liver weights were also linearly (P < 0.05) increased if pigs were fed diets containing CMB, but kidney weights were linearly (P < 0.05) decreased by the addition of CM-CV to the diets. Thyroid gland weights increased linearly (P < 0.05) for pigs fed diets containing CMA. No differences were observed in heart and bone weights if CM was added to the diets. Addition of any of the 3 CM linearly (P < 0.05) increased bone ash percentage in the metacarpals. Inclusion of CMA or CM-CV linearly (P < 0.05) decreased concentrations of serum triiodothyronine, and the inclusion of CMA also linearly (P < 0.05) decreased serum thyroxine concentrations. No differences were observed for complete blood counts or blood urea nitrogen if CM was added to the diets. In conclusion, up to 20% high-protein CM or CM-CV may be included in diets for weanling pigs from 2 wk postweaning without reducing growth performance or negatively affecting organ, bone, or blood parameters. In some instances, it may also be possible to use greater inclusion rates.
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Cholecalciferol, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Retinyl acetate, BioReagent, solid or viscous liquid, synthetic, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Supelco
Retinol solution, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80