MilliporeSigma
  • Micellar-enhanced ultrafiltration of copper ions using sodium dodecyl sulfate and its mixture with Brij 35, Tween 80 and Triton X-100.
Micellar-enhanced ultrafiltration of copper ions using sodium dodecyl sulfate and its mixture with Brij 35, Tween 80 and Triton X-100.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-05-17)
Baowei Zhao, Ruirui Li, Jinkui Zhong, Li Zhang
ABSTRACT
The performance of copper ion removal using sodium dodecyl sulfate (SDS) and its mixtures with Brij 35, Tween 80 (TW80) and Triton X-100 (TX100) by micellar-enhanced ultrafiltration (MEUF) was investigated. The effects of the molar ratio of nonionic surfactant to SDS on the critical micelle concentration (CMC) of SDS/Brij 35, SDS/TW80 and SDS/TX100, the removal efficiency of Cu(2+), the residual concentration of SDS in the permeate solution and the permeate flux were tested. The results showed that the CMCs of the mixed surfactants were sharply less than that of pure SDS. The removal efficiencies of Cu(2+) were up to the maximum values 98.3 and 95.8% when the molar ratios of Brij 35 and TW80 to SDS were 0.3, and it was 93.5% given 0.7 molar ratio of TX100 to SDS. The concentration of SDS in the permeate decreased dramatically with the addition of nonionic surfactant, and the permeate flux decreased slightly as the molar ratio increased. Compared with the performance by single SDS, the mixed SDS/Brij 35, SDS/TW80 and SDS/TX100 at an optimum composition could result in not only higher rejection of Cu(2+) but also much less dosage of surfactant and concentration of SDS in the permeate.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Triton X-100 solution, BioUltra, for molecular biology, ~10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Triton X-102
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Octoxinol 10, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Triton X-45
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, for molecular biology