MilliporeSigma
  • Reduction of MOPEG levels in cerebrospinal fluid of psychotic women after electroconvulsive treatment.
Reduction of MOPEG levels in cerebrospinal fluid of psychotic women after electroconvulsive treatment.
Psychopharmacology (1979-08-08)
C Härnryd, L Bjerkenstedt, V E Grimm, G Sedvall
PMID115032
ABSTRACT
The effect of ECT on concentrations of monoamine metabolites in lumbar CSF of psychotic women with a schizophrenic symptomatology was examined. After a series of ECT there was a significant reduction of the concentration of the major noradrenaline metabolite, MOPEG. Levels of HVA, 5-HIAA, prolactin, or total protein in CSF were not significantly influenced by treatment. The results indicate a specific alteration of central noradrenaline metabolism in relation to ECT.
Product Description
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Alanine, ≥99%
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
SAFC
L-Threonine
SAFC
Glycine
SAFC
L-Tyrosine
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)