MilliporeSigma
  • Flow, packing and compaction properties of novel coprocessed multifunctional directly compressible excipients prepared from tapioca starch and mannitol.
Flow, packing and compaction properties of novel coprocessed multifunctional directly compressible excipients prepared from tapioca starch and mannitol.
Pharmaceutical development and technology (2013-10-05)
Oluwatomide Adeoye, Gbenga Alebiowu
ABSTRACT
Novel multifunctional excipients were prepared by coprocessing tapioca starch with mannitol using two methods viz; co-grinding and co-fusion. The flow, packing and compaction properties of the native and novel excipients were evaluated by using density, Hausner's ratio, angle of repose, the maximum volume reduction, consolidation index, the rate of consolidation, angle of internal friction, morphological properties, Heckel analysis, tensile strength and dilution potential as evaluation parameters. The study revealed that the method of coprocessing, particle size and particle shape influenced the properties of the resulting novel excipients. Co-grinding was less effective than co-fusion in the preparation of excipients with enhanced properties. The study concluded that coprocessing tapioca starch and mannitol will enhance the flow, packing and compaction properties of the novel excipient and that the co-fusion method of coprocessing would produce novel excipients with enhanced direct compression potential compared to the co-grinding method.
Sigma-Aldrich
Starch from corn, practical grade
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, SAJ first grade, ≥99.0%
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Starch from potato, Soluble
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Starch from wheat, Unmodified
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, SAJ special grade, ≥99.0%
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard