MilliporeSigma
  • Fibroblast growth factor (Fgf) 21 is a novel target gene of the aryl hydrocarbon receptor (AhR).
Fibroblast growth factor (Fgf) 21 is a novel target gene of the aryl hydrocarbon receptor (AhR).
Toxicology and applied pharmacology (2014-04-29)
Xingguo Cheng, Saurabh G Vispute, Jie Liu, Christine Cheng, Alexei Kharitonenkov, Curtis D Klaassen
ABSTRACT
The toxic effects of dioxins, such as 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD), mainly through activation of the aryl hydrocarbon receptor (AhR) are well documented. Fibroblast growth factor (Fgf) 21 plays critical roles in metabolic adaptation to fasting by increasing lipid oxidation and ketogenesis in the liver. The present study was performed to determine whether activation of the AhR induces Fgf21 expression. In mouse liver, TCDD increased Fgf21 mRNA in both dose- and time-dependent manners. In addition, TCDD markedly increased Fgf21 mRNA expression in cultured mouse and human hepatocytes. Moreover, TCDD increased mRNA (in liver) and protein levels (in both liver and serum) of Fgf21 in wild-type mice, but not in AhR-null mice. Chromatin immunoprecipitation assays showed that TCDD increased AhR protein binding to the Fgf21 promoter (-105/+1 base pair). Fgf21-null mice administered 200μg/kg of TCDD died within 20days, whereas wild-type mice receiving the same treatment were still alive at one month after administration. This indicates that TCDD-induced Fgf21 expression protects against TCDD toxicity. Diethylhexylphthalate (DEHP) pretreatment attenuated TCDD-induced Fgf21 expression in mouse liver and white adipose tissue, which may explain a previous report that DEHP pretreatment decreases TCDD-induced wasting. In conclusion, Fgf21 appears to be a target gene of AhR-signaling pathway in mouse and human liver.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
SKU
Pack Size
Availability
Price
Quantity
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
SKU
Pack Size
Availability
Price
Quantity
Supelco
Chloroform, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MOPS, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL