MilliporeSigma
  • Severe neutrophil-dominated inflammation and enhanced myelopoiesis in IL-33-overexpressing CMV/IL33 mice.
Severe neutrophil-dominated inflammation and enhanced myelopoiesis in IL-33-overexpressing CMV/IL33 mice.
Journal of immunology (Baltimore, Md. : 1950) (2014-12-17)
Dominique Talabot-Ayer, Praxedis Martin, Christian Vesin, Christian Alexander Seemayer, Solenne Vigne, Cem Gabay, Gaby Palmer
ABSTRACT
IL-33 is a cytokine of the IL-1 family, which signals through the ST2 receptor. Previous studies emphasized a role for IL-33 in shaping innate and adaptive immune responses. IL-33 was also reported to modulate myelopoiesis and myeloid cell activity. In this article, we describe IL-33-overexpressing CMV/IL33 and LysM/IL33 mice, which display an inflammatory phenotype associated with growth retardation and paw swelling. The phenotype of CMV/IL33 mice is dependent on activation of the ST2 receptor and is characterized by extensive neutrophil infiltration into different organs, including the paws. Local or systemic levels of proinflammatory mediators such as IL-1β, Cxcl-1, G-CSF, and IL-6 are increased. CMV/IL-33 mice also suffer from anemia, thrombocytosis, and a marked dysregulation of myelopoiesis, leading to an important increase in myeloid cell production or accumulation in bone marrow (BM), spleen, and peripheral blood. Consistently, recombinant IL-33 induced proliferation of myeloid lineage cells in BM-derived granulocyte cultures, whereas IL-33 knockout mice exhibited minor deficiencies in spleen and BM myeloid cell populations. Our observations reveal a neutrophil-dominated inflammatory phenotype in IL-33-overexpressing CMV/IL33 and LysM/IL33 mice, and highlight important regulatory effects of IL-33 on myelopoiesis in vitro and in vivo, where excessive IL-33 signaling can translate into the occurrence of a myeloproliferative disorder.
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M