MilliporeSigma
  • The Aβ peptides-activated calcium-sensing receptor stimulates the production and secretion of vascular endothelial growth factor-A by normoxic adult human cortical astrocytes.
The Aβ peptides-activated calcium-sensing receptor stimulates the production and secretion of vascular endothelial growth factor-A by normoxic adult human cortical astrocytes.
Neuromolecular medicine (2014-06-21)
Ilaria Dal Prà, Ubaldo Armato, Franco Chioffi, Raffaella Pacchiana, James F Whitfield, Balu Chakravarthy, Li Gui, Anna Chiarini
ABSTRACT
The excess vascular endothelial growth factor (VEGF) produced in the Alzheimer's disease (AD) brain can harm neurons, blood vessels, and other components of the neurovascular units (NVUs). But could astrocytes partaking in networks of astrocyte-neuron teams and connected to blood vessels of NVUs contribute to VEGF production? We have shown with cultured cerebral cortical normal (i.e., untransformed) adult human astrocytes (NAHAs) that exogenous amyloid-β peptides (Aβs) stimulate the astrocytes to make and secrete large amounts of Aβs and nitric oxide by a mechanism mediated through the calcium-sensing receptor (CaSR). Here, we report that exogenous Aβs stimulate the NAHAs to produce and secrete even VEGF-A through a CaSR-mediated mechanism. This is indicated by the ability of Aβs to specifically bind the CaSR, and the capability of a CaSR activator, the "calcimimetic" NPS R-568, to imitate, and of the CaSR antagonist, "calcilytic" NPS 2143, to inhibit, the Aβs stimulation of VEGF-A production and secretion by the NAHAs. Thus, Aβs that accumulate in the AD brain may make the astrocytes that envelop and functionally collaborate with neurons into multi-agent AD-driving "machines" via a CaSR signaling mechanism(s). These observations suggest the possibility that CaSR allosteric antagonists such as NPS 2143 might impede AD progression.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
SAFC
BIS-TRIS
Sigma-Aldrich
Anti-LMNB1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard