MilliporeSigma
  • Molecular and biochemical evidences on the protective effects of triiodothyronine against phosphine-induced cardiac and mitochondrial toxicity.
Molecular and biochemical evidences on the protective effects of triiodothyronine against phosphine-induced cardiac and mitochondrial toxicity.
Life sciences (2015-08-05)
Amir Hossein Abdolghaffari, Amir Baghaei, Reza Solgi, Maziar Gooshe, Maryam Baeeri, Mona Navaei-Nigjeh, Shokoufeh Hassani, Abbas Jafari, Seyed Mehdi Rezayat, Ahmad Reza Dehpour, Shahram Ejtemaei Mehr, Mohammad Abdollahi
ABSTRACT
Aluminum phosphide (AlP) is a widely used fumigant and rodenticide. While AlP ingestion leads to high mortality, its exact mechanism of action is unclear. There are ample evidences suggesting cardioprotective effects of triiodothyronine (T3). In this study, we aimed to examine the potential of T3 in the protection of a rat model of AlP induced cardiotoxicity. In order to induce AlP intoxication animals were intoxicated with AlP (12 mg/kg; LD50) by gavage. In treatment groups, T3 (1, 2 and 3 μg/kg) was administered intra-peritoneally 30 min after AlP administration. Animals were connected to the electronic cardiovascular monitoring device simultaneously after T3 administration. Then, electrocardiogram (ECG), blood pressure (BP), and heart rate (HR) were monitored for 180 min. Additionally, 24h after AlP intoxication, rats were deceased and the hearts were dissected out for evaluation of oxidative stress, cardiac mitochondrial function (complexes I, II and IV), ATP/ADP ratio, caspases 3 & 9, and apoptosis by flow cytometry. The results demonstrated that AlP intoxication causes cardiac toxicity presenting with changes in ECG patterns such as decrement of HR, BP and abnormal QRS complexes, QTc and ST height. T3 at a dose of 3 μg/kg significantly improved ECG and also oxidative stress parameters. Furthermore, T3 administration could increase mitochondrial function and ATP levels within the cardiac cells. In addition, administration of T3 showed a reduction in apoptosis through diminishing the caspase activities and improving cell viability. Overall, the present data demonstrate the beneficial effects of T3 in cardiotoxicity of AlP.
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O