MilliporeSigma
  • 5-demethylnobiletin promotes the formation of polymerized tubulin, leads to G2/M phase arrest and induces autophagy via JNK activation in human lung cancer cells.
5-demethylnobiletin promotes the formation of polymerized tubulin, leads to G2/M phase arrest and induces autophagy via JNK activation in human lung cancer cells.
The Journal of nutritional biochemistry (2015-03-15)
Yu-Kuo Chen, Hsin-Chieh Wang, Chi-Tang Ho, Hisn-Yu Chen, Shiming Li, Hong-Lin Chan, Ting-Wen Chung, Kok-Tong Tan, Yi-Ron Li, Chi-Chen Lin
ABSTRACT
5-Demethylnobiletin is a hydroxylated polymethoxyflavone found in citrus plants that shows antiproliferative activities in several cancer cell lines. In this study, we investigated the effects and underlying molecular mechanisms of 5-demethylnobiletin on inhibition of cell growth, apoptosis, cell cycle and autophagy in A549 and CL1-5 lung cancer cells. The results of the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay suggested that 5-demethylnobiletin inhibited cell growth in a dose- and time-dependent manner. Flow cytometry results suggested that 5-demethylnobiletin inhibited proliferation in lung cancer cells by inducing G2/M cell cycle phase arrest but predominantly not through apoptosis. Western blot results illustrated that the blockade of the cell cycle was associated with reduced levels of cdc25 and cdc2. Notably, our results indicated that 5-demethylnobiletin induced significant abnormal microtubule dynamics in A549 and CL1-5 cells, a novel finding. Studies conducted with isolated tubulin and docking models suggest that 5-demethylnobiletin promoted the polymerization of microtubules and bound to the taxol site. Additionally, 5-demethylnobiletin might also induce autophagy via activation of the JNK signaling pathway in A549 and CL1-5 cells. Pretreatment of the cells with the autophagy inhibitor 3-methyladenine significantly potentiated 5-demethylnobiletin-induced apoptosis, suggesting that 5-demethylnobiletin-induced autophagy mitigated cell apoptosis. Further investigation revealed that 5-demethylnobiletin inhibition of CL1-5 lung cancer cell growth was reproducible in a nude mouse model. Taken together, these studies suggest that 5-demethylnobiletin has anti-lung cancer efficacy both in vitro and in vivo possibly through induction of G2/M arrest, autophagy and apoptosis.
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glyceryl trioctanoate, ≥95% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)