MilliporeSigma
  • Ultrasound molecular imaging of tumor angiogenesis with a neuropilin-1-targeted microbubble.
Ultrasound molecular imaging of tumor angiogenesis with a neuropilin-1-targeted microbubble.
Biomaterials (2015-05-03)
Hua Zhang, Sarah Tam, Elizabeth S Ingham, Lisa M Mahakian, Chun-Yen Lai, Spencer K Tumbale, Tambet Teesalu, Neil E Hubbard, Alexander D Borowsky, Katherine W Ferrara
ABSTRACT
Ultrasound molecular imaging has great potential to impact early disease diagnosis, evaluation of disease progression and the development of target-specific therapy. In this paper, two neuropilin-1 (NRP) targeted peptides, CRPPR and ATWLPPR, were conjugated onto the surface of lipid microbubbles (MBs) to evaluate molecular imaging of tumor angiogenesis in a breast cancer model. Development of a molecular imaging agent using CRPPR has particular importance due to the previously demonstrated internalizing capability of this and similar ligands. In vitro, CRPPR MBs bound to an NRP-expressing cell line 2.6 and 15.6 times more than ATWLPPR MBs and non-targeted (NT) MBs, respectively, and the binding was inhibited by pretreating the cells with an NRP antibody. In vivo, the backscattered intensity within the tumor, relative to nearby vasculature, increased over time during the ∼6 min circulation of the CRPPR-targeted contrast agents providing high contrast images of angiogenic tumors. Approximately 67% of the initial signal from CRPPR MBs remained bound after the majority of circulating MBs had cleared (8 min), 8 and 4.5 times greater than ATWLPPR and NT MBs, respectively. Finally, at 7-21 days after the first injection, we found that CRPPR MBs cleared faster from circulation and tumor accumulation was reduced likely due to a complement-mediated recognition of the targeted microbubble and a decrease in angiogenic vasculature, respectively. In summary, we find that CRPPR MBs specifically bind to NRP-expressing cells and provide an effective new agent for molecular imaging of angiogenesis.
Sigma-Aldrich
Chloroform, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Glycerol, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Glycerol, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15000000 units
Estimated to ship on March 22, 2025
$200.00
75000000 units
Estimated to ship on March 22, 2025
$687.00
150000000 units
Estimated to ship on March 22, 2025
$1,170.00