MilliporeSigma
  • Antiherpes Activity and Skin/Mucosa Distribution of Flavonoids from Achyrocline satureioides Extract Incorporated into Topical Nanoemulsions.
Antiherpes Activity and Skin/Mucosa Distribution of Flavonoids from Achyrocline satureioides Extract Incorporated into Topical Nanoemulsions.
BioMed research international (2015-06-24)
Juliana Bidone, Débora Fretes Argenta, Jadel Kratz, Letícia Ferreira Pettenuzzo, Ana Paula Horn, Letícia Scherer Koester, Valquíria Linck Bassani, Claudia Maria Oliveira Simões, Helder Ferreira Teixeira
ABSTRACT
This study investigated the inhibitory effects of Achyrocline satureioides extract (ASE) incorporated into a topical nanoemulsion on Herpes Simplex Virus type 1 (HSV-1/KOS strain) replication, as well as the distribution of the main ASE flavonoids (quercetin, luteolin, and 3-O-methylquercetin) in porcine skin and mucosa. The ASE-loaded nanoemulsion showed more pronounced effects against HSV-1 replication when compared to the ASE or pure quercetin, as determined by the viral plaque number reduction assay. All flavonoids were detected in the skin epidermis (2.2 µg/cm(2)) and the mucosa upper layers (3.0 µg/cm(2)) from ASE-loaded nanoemulsion until 8 h after topical application. A higher amount of flavonoids was detected when these tissues were impaired, especially in deeper mucosa layers (up to 7-fold). Flavonoids were detected in the receptor fluid only when the mucosa was injured. Such results were supported by confocal microscopy images. Overall, these findings suggest that the tested ASE-loaded nanoemulsion has potential to be used topically for herpes infections.
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis