MilliporeSigma
  • Imidazolium salts as small-molecule urinary bladder exfoliants in a murine model.
Imidazolium salts as small-molecule urinary bladder exfoliants in a murine model.
Antimicrobial agents and chemotherapy (2015-07-01)
Patrick O Wagers, Kristin M Tiemann, Kerri L Shelton, William G Kofron, Matthew J Panzner, Karen L Wooley, Wiley J Youngs, David A Hunstad
ABSTRACT
We present a novel family of small-molecule urinary bladder exfoliants that are expected to be of great value in preclinical studies of urologic conditions and have improved potential for translation compared with prior agents. There is broad urologic interest in the therapeutic potential of such exfoliating agents. The primary agent used in preclinical models, the cationic peptide protamine sulfate (PS), has limited translational potential due to concerns including systemic adverse reactions and bladder tissue injury. Intravesical application of a safe, systemically nontoxic exfoliant would have potential utility in the eradication of Escherichia coli and other uropathogens that reside in the bladder epithelium following cystitis, as well as in chronic bladder pain and bladder cancer. Here, we introduce a family of imidazolium salts with potent and focused exfoliating activity on the bladder epithelium. Synthesis and purification were straightforward and scalable, and the compounds exhibited prolonged stability in lyophilized form. Most members of the compound family were cytotoxic to cultured uroepithelial cells, with >10-fold differences in potency across the series. Upon topical (intravesical) administration of selected compounds to the murine bladder, complete epithelial exfoliation was achieved with physiologically relevant imidazolium concentrations and brief contact times. The exfoliative activity of these compounds was markedly improved in comparison to PS, as assessed by microscopy, immunofluorescence, and immunoblotting for uroplakins. Bladder uroepithelium regenerated within days to yield a histologically normal appearance, and no toxicity was observed. Finally, the chemical scaffold offers an opportunity for inclusion of antimicrobials or conjugation with chemotherapeutic or other moieties.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%