Sigma-Aldrich
Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Hemoglobin from bovine blood, lyophilized powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Monellin from Dioscoreophyllum cumminsii (serendipity berry)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
α-Chymotrypsinogen A from bovine pancreas, essentially salt-free, lyophilized powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Papain from papaya latex, buffered aqueous suspension, 2× Crystallized, ≥16 units/mg protein
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type I-B, lyophilized powder, ≥50,000 units/mg solid
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Citrate Synthase from porcine heart, ammonium sulfate suspension, ≥100 units/mg protein
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ubiquitin from bovine erythrocytes, BioUltra, ≥98% (SDS-PAGE), essentially salt-free, lyophilized powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Trypsin-chymotrypsin inhibitor from Glycine max (soybean), lyophilized powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Thaumatin from Thaumatococcus daniellii
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Trypsinogen from bovine pancreas, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein (E1%/280, after activation to trypsin)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
3-Phosphoglyceric Phosphokinase from baker′s yeast (S. cerevisiae), ammonium sulfate suspension, ≥500 units/mg protein
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triosephosphate Isomerase from baker′s yeast (S. cerevisiae), Type I, ammonium sulfate suspension, ~10,000 units/mg protein
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutathione S-Transferase from equine liver, lyophilized powder, ≥25 units/mg protein
Sign Into View Organizational & Contract Pricing