MilliporeSigma
  • Cytotoxicity testing of silver-containing burn treatments using primary and immortal skin cells.
Cytotoxicity testing of silver-containing burn treatments using primary and immortal skin cells.
Burns : journal of the International Society for Burn Injuries (2014-04-29)
Benjawan Boonkaew, Margit Kempf, Roy Kimble, Leila Cuttle
ABSTRACT
A novel burn wound hydrogel dressing has been previously developed which is composed of 2-acrylamido-2-methylpropane sulfonic acid sodium salt with silver nanoparticles (silver AMPS). This study compared the cytotoxicity of this dressing to the commercially available silver products; Acticoat™, PolyMem Silver(®) and Flamazine™ cream. Human keratinocytes (HaCaT and primary HEK) and normal human fibroblasts (NHF) were exposed to dressings incubated on Nunc™ polycarbonate inserts for 24, 48 and 72h. Four different cytotoxicity assays were performed including; Trypan Blue cell count, MTT, Celltiter-Blue™ and Toluidine Blue surface area assays. The results were expressed as relative cell viability compared to an untreated control. The cytotoxic effects of Acticoat™ and Flamazine™ cream were dependent on exposure time and cell type. After 24h exposure, Acticoat™ and Flamazine™ cream were toxic to all tested cell lines. Surprisingly, HaCaTs treated with Acticoat™ and Flamazine™ had an improved ability to survive at 48 and 72h while HEKs and NHFs had no improvement in survival with any treatment. The novel silver hydrogel and PolyMem Silver(®) showed low cytotoxicity to all tested cell lines at every time interval and these results support the possibility of using the novel silver hydrogel as a burn wound dressing. Researchers who rely on HaCaT cells as an accurate keratinocyte model should be aware that they can respond differently to primary skin cells.
Sigma-Aldrich
1-Hexadecanol, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
1-Hexadecanol, 95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
1-Hexadecanol, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
Cetyl alcohol, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
1,2-Propanediol, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
USP
Cetyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
Cetyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Cetyl alcohol, SAJ special grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00