MilliporeSigma
  • A simple and low-cost platform technology for producing pexiganan antimicrobial peptide in E. coli.
A simple and low-cost platform technology for producing pexiganan antimicrobial peptide in E. coli.
Biotechnology and bioengineering (2014-11-27)
Chun-Xia Zhao, Mirjana Dimitrijev Dwyer, Alice Lei Yu, Yang Wu, Sheng Fang, Anton P J Middelberg
ABSTRACT
Antimicrobial peptides, as a new class of antibiotics, have generated tremendous interest as potential alternatives to classical antibiotics. However, the large-scale production of antimicrobial peptides remains a significant challenge. This paper reports a simple and low-cost chromatography-free platform technology for producing antimicrobial peptides in Escherichia coli (E. coli). A fusion protein comprising a variant of the helical biosurfactant protein DAMP4 and the known antimicrobial peptide pexiganan is designed by joining the two polypeptides, at the DNA level, via an acid-sensitive cleavage site. The resulting DAMP4(var)-pexiganan fusion protein expresses at high level and solubility in recombinant E. coli, and a simple heat-purification method was applied to disrupt cells and deliver high-purity DAMP4(var)-pexiganan protein. Simple acid cleavage successfully separated the DAMP4 variant protein and the antimicrobial peptide. Antimicrobial activity tests confirmed that the bio-produced antimicrobial peptide has the same antimicrobial activity as the equivalent product made by conventional chemical peptide synthesis. This simple and low-cost platform technology can be easily adapted to produce other valuable peptide products, and opens a new manufacturing approach for producing antimicrobial peptides at large scale using the tools and approaches of biochemical engineering.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Tricine, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Tricine, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
SAFC
Tricine
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%