MilliporeSigma
  • Aromatase controls Sjögren syndrome-like lesions through monocyte chemotactic protein-1 in target organ and adipose tissue-associated macrophages.
Aromatase controls Sjögren syndrome-like lesions through monocyte chemotactic protein-1 in target organ and adipose tissue-associated macrophages.
The American journal of pathology (2014-12-03)
Akihiko Iwasa, Rieko Arakaki, Naoko Honma, Aya Ushio, Akiko Yamada, Tomoyuki Kondo, Emi Kurosawa, Satoko Kujiraoka, Takaaki Tsunematsu, Yasusei Kudo, Eiji Tanaka, Noriko Yoshimura, Nobuhiro Harada, Yoshio Hayashi, Naozumi Ishimaru
ABSTRACT
Several autoimmune diseases are known to develop in postmenopausal women. However, the mechanism by which estrogen deficiency influences autoimmunity is unknown. Aromatase is an enzyme that converts androgens to estrogens. Herein, we used female aromatase gene knockout (ArKO) mice as a model of estrogen deficiency to investigate the molecular mechanism that underlies the onset and development of autoimmunity. Histological analyses showed that inflammatory lesions in the lacrimal and salivary glands of ArKO mice increased with age. Adoptive transfer of spleen cells or bone marrow cells from ArKO mice into recombination activating gene 2 knockout mice failed to induce the autoimmune lesions. Expression of mRNA encoding proinflammatory cytokines and monocyte chemotactic protein-1 increased in white adipose tissue of ArKO mice and was significantly higher than that in wild-type mice. Moreover, an increased number of inflammatory M1 macrophages was observed in white adipose tissue of ArKO mice. A significantly increased monocyte chemotactic protein-1 mRNA expression of the salivary gland tissue in ArKO was found together with adiposity. Furthermore, the autoimmune lesions in a murine model of Sjögren syndrome were exacerbated by administration of an aromatase inhibitor. These results suggest that aromatase may play a key role in the pathogenesis of Sjögren syndrome-like lesions by controlling the target organ and adipose tissue-associated macrophage.
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material