Sigma-Aldrich
Chloroform, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Supelco
Chloroform, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Pioglitazone for system suitability, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets